Learn More
Alfa Aesar™ Caspase-3 Inhibitor III
Manufacturer:  Alfa Aesar™ J64442LB0
 View more versions of this product
Catalog No. AAJ64442LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
-30°C to -10°C
Caspase-3 Inhibitor
285570-60-7
550.94
Soluble in DMSO
Off-white to brown
Powder
A potent, cell-permeable, and irreversible inhibitor of caspase-3; also inhibits caspases -6, -7, -8, and -10. Commonly used at concentrations up to 100 →M to examine the role of caspase-3-dependent apoptosis
C21H31ClN4O11
Ac-DEVD-CMK; N-Acetyl-Asp-Glu-Val-Asp chloromethyl ketone