Learn More
Caspase-3 Mouse anti-Human, Mouse, Rat, Alexa Fluor 700, Clone: CPP32 4-1-18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB500210AF700
Catalog No. NBV210AF700
Add to cart
Description
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
Specifications
Caspase-3
CPP32 4-1-18
Western Blot, Immunohistochemistry-Paraffin
Purified
CASP3
Full-length recombinant human Caspase 3 [UniProt# P42574]
Protein G purified
Alzheimers Research, Apoptosis, Cancer, Caspases, Neurodegeneration, Neuroscience, Phospho Specific
Primary
836
Western Blot, Immunohistochemistry (Paraffin)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Mouse
IgG2a Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat