Learn More
Enzo Life Sciences Caspase 3 polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP113F
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Caspase-3 protein (full length). UniProt: P42574. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Yeast, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-658-751