Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Rabbit anti-Human,Mouse,Rat,Bovine, KHL, Clone: 3J16, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Caspase-3
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB122125MI
View Documents
MilliporeSigma
ZRB122125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12214XMI
View Documents
MilliporeSigma
ZRB12214X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human,Mouse,Rat,Bovine samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Caspase-3
KHL
Rabbit
CASP-3;Apopain;Cysteine protease CPP32;CPP-32;EC:3.4.22.56;Protein Yama;SREBP cleavage activity 1;SCA-1
KLH-conjugated linear peptide corresponding to 12 amino acids from the p17 subunit of human Caspase-3.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat, Bovine
CASP3, CPP32
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit