Learn More
Enzo Life Sciences Caspase 8 monoclonal antibody (5F7) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM118E
Encompass_Preferred
Clone: 5F7. Host: Mouse. Isotype: IgG2b. Immunogen: C-terminal fragment of recombinant human caspase-8 (amino acids 176-460). UniProt: Q14790. Species Reactivity: Human. Applications: IHC (PS), IP, WB. Recommended dilutions: Immunoprecipitation (1:50). Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-658-712