Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Caspase 9/LAP6 Ab-4, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments.

Specifications

Antigen Caspase 9/LAP6 Ab-4
Concentration 1mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Format Concentrated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB1205P1
View Documents
Thermo Scientific™
RB1205P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Caspase 9/LAP6 Ab-4, Rabbit Polyclonal Antibody. 

 

Activation of procaspase-9 by Apaf-1 in the cytochrome c/dATP-dependent pathway requires proteolytic cleavage to generate the mature caspase molecule. Deletion of the Apaf-1 WD-40 repeats makes Apaf-1 constitutively active and capable of processing procaspase-9 independent of cytochrome c and ATP. Apaf-1-mediated processing of procaspase-9 occurs at Asp-315 by an intrinsic autocatalytic activity of procaspase-9 itself. Apaf-1 can form oligomers and may facilitate procaspase-9 autoactivation by oligomerizing its precursor molecules. Once activated, caspase-9 can initiate a caspase cascade involving the downstream executioners caspase-3, -6, and -7.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow and Sheep. Others not tested.

Epitope: aa 1-134

Immunogen: Recombinant protein encoding aa 1-134 of human caspase 9

Molecular Weight: 46-48 kDa

Positive Control: Jurkat cells, Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Caspase 9/LAP6 Ab-4
Immunoprecipitation, Western Blot
Concentrated
RUO
Primary
IgG
1mg/mL
Polyclonal
Rabbit
Apoptosis
Recombinant protein encoding aa 1-134 of human caspase 9
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit