Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASS4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310642100UL
Description
CASS4 Polyclonal specifically detects CASS4 in Human samples. It is validated for Western Blot.Specifications
CASS4 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
C20orf32, Cas scaffolding protein family member 4, HEF1-EFS-p130Cas-like protein, HEFLchromosome 20 open reading frame 32, HEF-like protein, HEPLCAS4 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human CASS4. Peptide sequence ALLARALYDNCPDCSDELAFSRGDILTILEQHVPESEGWWKCLLHGRQGL | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
57091 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction