Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Cassette, Sklar, 7 x 3-1/2 x 6/7 in. Instrument Cassette Medium with Hinged Lid and Silicone Instrument Holders, Holds Max. 7 Instruments. Sklar's Instrument Cassettes are used for cleaning, storage and sterilization. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106506
Instrument Cassette, Sklar, 7 x 3-1/2 x 6/7 in. Instrument Cassette Medium with Hinged Lid and Silicone Instrument Holders, Holds Max. 7 Instruments. Sklar's Instrument Cassettes are used for cleaning, storage and sterilization. The silicone supports are repositionable to assist in arranging various instruments. The cassettes are available with a hinged or detachable lid. This product is a medium cassette with a hinged lid and can hold approximately 7 instruments.
Catalog No. 50-143-7223