Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Cassette, Sklar, 7 x 2-1/3 x 6/7 in. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106525
Instrument Cassette, Sklar, 7 x 2-1/3 x 6/7 in. Instrument Cassette Open Clip-in with moveable Silicone Instrument Holders, Holds Max. 5 Instruments. Sklar Instrument Cassettes are used for cleaning, storing, and sterilizing instruments.
Catalog No. 50-143-7230