Learn More
Ihc World LLCSupplier Diversity Partner CASSETTE STORAGE DFS Item
Manufacturer:  Ihc World LLCSupplier Diversity Partner M4957
Made of durable, waterproof, heavy duty cardboard for long term storage. Each box can store up to 165 embedding rings or 250 cassettes per drawer.
Catalog No. NC0152522