Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Cassette, Sklar, 15 x 8-1/4 x 1-1/2 in. Instrument Cassette X-Large with Detachable Lid, Open Configuration. Sklar Instrument Cassettes are used for cleaning, storing, and sterilizing instruments. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106521
Instrument Cassette, Sklar, 15 x 8-1/4 x 1-1/2 in. Instrument Cassette X-Large with Detachable Lid, Open Configuration. Sklar Instrument Cassettes are used for cleaning, storing, and sterilizing instruments. The silicone supports are repositionable to assist in arranging and stabilizing instruments. Cassettes are available with a hinged or detachable lid.
Catalog No. 50-143-7229