Learn More
Sklar InstrumentsSupplier Diversity Partner Silicone Instrument Holder for Cassettes with Hinged Lid, Sklar, 2 1/3 in. Silicone Instrument Holder for Cassettes with Hinged Lid 3 Piece, Maximum 4 Instrument Configuration, Blue. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106545
Silicone Instrument Holder for Cassettes with Hinged Lid, Sklar, 2 1/3 in. Silicone Instrument Holder for Cassettes with Hinged Lid 3 Piece, Maximum 4 Instrument Configuration, Blue. Sklar's Silicone Instrument Holders are used for positioning instruments inside a cassette with a hinged lid used for cleaning, storage, or sterilization. The holders are repositionable to assist in storing different types and size instruments within a casette. They are available in a variety of colors and sold in sets of 3. This product is blue, has an overall length of 2-1/3 inches and holds a maximun of 4 instruments.
Catalog No. 50-143-7245