Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Castanospermine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

79831-76-8
C8H15NO4
189.211
JDVVGAQPNNXQDW-TVNFTVLESA-N
[1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
54445
CHEBI:27860

Glucosidases α and β inhibitor

Potent inhibitor of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids). Inhibits HIV syncytium formation and replication.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C1CN2CC(C(C(C2C1O)O)O)O
IUPAC Name (1S,6S,7R,8R,8aR)-1,2,3,5,6,7,8,8a-octahydroindolizine-1,6,7,8-tetrol
Formula Weight 189.21
Chemical Name or Material Castanospermine
Grade >98%

Recommended Storage : Store at Room Temperature