Learn More
Castor Oil, FCC, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Drum
Steel Pail
C1108, 8001-79-4
Spectrum™ Castor Oil, FCC is used as mold inhibitor for packaging and in flavorings and candy. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Steel Pail