Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Castor Oil, Sulfated, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
8002-33-3
8002-33-3

CA209, 8002-33-3

Spectrum™ Castor Oil, Sulfated is an oil that is derived from the castor beans and is made by adding sulfuric acid to the oil. A colorless oil that has no taste or smell and boils at 313 C, it is used as a synthetic detergent in formulating lubricants, softeners, and dyeing assistants. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Quantity 20 L
Water 24-32%
Packaging Poly Drum
CAS Max % 100%