Learn More
Sklar InstrumentsSupplier Diversity Partner TC Castroviejo Needle Holder, Sklar, 5-1/2 in., Curved, Cross-Serrated, Flat Thumb, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218047
TC Castroviejo Needle Holder, Sklar, 5-1/2 in., Curved, Cross-Serrated, Flat Thumb, Tungsten Carbide / Stainless Steel. Sklar's Tungsten Carbide Castroviejo Needle Holder is a multipurpose microsurgical instrument that is often used in ophthalmic and plastic surgeries. It features a double spring, flat handle, and it is available with straight or curved jaws. It is also available with or without a locking mechanism. The small tips of this needle holder are ideal for handling small, delicate needles and sutures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product has curved, cross-serrated jaws, an overall length of 5-1/2 inches and a locking mechanism.
Catalog No. 50-143-7733