Learn More
Aviva Systems Biology Corp CAT Antibody - middle region (ARP58437_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58437_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CAT. Alternate names: MGC138422, MGC138424ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-62307