Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen CAT1
Immunogen Synthetic peptides corresponding to SLC7A1(solute carrier family 7 (cationic amino acid transporter, y+ system), member 1) The peptide sequence was selected from the N terminal of SLC7A1. Peptide sequence ELWAFITGWNLILSYIIGTSSVARAWSATFDELI
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15983620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15983620UL
20ul Each for $149.00
Add to cart
 
NBP159836 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159836
100 ul Each for $329.00
Add to cart
 
Description

Description

CAT1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

CAT1
Rabbit
Polyclonal
Unconjugated
6541
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to SLC7A1(solute carrier family 7 (cationic amino acid transporter, y+ system), member 1) The peptide sequence was selected from the N terminal of SLC7A1. Peptide sequence ELWAFITGWNLILSYIIGTSSVARAWSATFDELI
Primary
IgG
amino acid transporter, cationic 1, ATRC1ERR, CAT-1System Y+ basic amino acid transporter, Ecotropic retroviral leukemia receptor homolog, ecotropic retroviral receptor, Ecotropic retrovirus receptor homolog, HCAT1, high affinity cationic amino acid transporter 1, REC1LCAT1, solute carrier family 7 (cationic amino acid transporter, y+ system), member 1, Solute carrier family 7 member 1
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit