Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catalase from Bovine Liver, Thermo Scientific Chemicals
Derived from Bovine liver
Supplier: Thermo Scientific J1288506
Description
- One unit of enzyme decomposes 1μmol H2O2/min at 25°C at pH 7.0
Specifications
≥3,000 U/mg | |
-20°C, Protect from moisture | |
5 g | |
Lyophilized |
7 | |
Bovine Liver | |
Catalase |
Safety and Handling
EINECSNumber : 232-577-1
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction