Learn More
MP Biomedicals™ Catalase from Human Erythrocytes (E.C.1.11.1.6.)
Negative for HBsAg and HIV antibodies
$687.00 - $2202.00
Specifications
Format | Liquid |
---|---|
Concentration | 1 mg/mL |
For Use With (Application) | Protecting cells against oxidative damage to proteins, lipids and nucleic acids |
Storage Buffer | Frozen solution of 1mg/mL in 1.5mM Sodium Phosphate, 20mM NaCl, pH 7.0 |
Content And Storage | -20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP219134150
|
MP Biomedicals™
0219134150 |
50 kU |
Each for $687.00
|
|
MP219134180
|
MP Biomedicals™
0219134180 |
100 kU |
Each for $1,095.00
|
|
MP219134183
|
MP Biomedicals™
0219134183 |
250 kU |
Each for $2,202.00
|
|
ICN19134125
|
MP Biomedicals™
0219134125 |
25 kU | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.
Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).
Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.
Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).
Specifications
Liquid | |
Protecting cells against oxidative damage to proteins, lipids and nucleic acids | |
-20°C | |
Catalase |
1 mg/mL | |
Frozen solution of 1mg/mL in 1.5mM Sodium Phosphate, 20mM NaCl, pH 7.0 | |
Human erythrocyte | |
256000 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.