Learn More
Catalase, Lyophilized, Spectrum™
C1328, 9001-05-2
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner C132810GMBL
 View more versions of this product
Catalog No. 18-601-515
Add to cart
Description
Description
Spectrum™ Catalase, Lyophilized , a tetramer of four polypeptide chains, is used in the food industry to remove hydrogen peroxide from milk in order to make cheese. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
Amber Glass Bottle