Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catalase Antibody (OTI1B8), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270342PEATT594
Description
Catalase Monoclonal specifically detects Catalase in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
Catalase | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
CAT | |
Full length recombinant protein of human Catalase (NP_111743) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG1 |
Flow Cytometry | |
OTI1B8 | |
Flow Cytometry | |
catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
Mouse | |
Affinity Purified | |
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
847.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction