Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase Mouse anti-Human, Monkey, PE/Cy5.5, Clone: OTI1B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270342PECY55

Catalog No. N270342PEC5


Add to cart

Description

Description

Catalase Monoclonal antibody specifically detects Catalase in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Catalase
OTI1B8
Flow Cytometry
Affinity Purified, Conjugated
CAT
Full length recombinant protein of human Catalase (NP_111743) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary
847.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
catalase, EC 1.11.1.6, MGC138422, MGC138424
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit