Learn More
Biomatik Corporation SYK (Phospho-Tyr348) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41079
The SYK (Phospho-Tyr348) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: SYK around the phosphorylation site of Tyr348. Applications: WB, IHC, ELISA. Gene ID: 6850. Gene Name: SYK. Alternative Names: Spleen tyrosine kinase; PROTEIN-TYROSINE KINASE SYK. Swiss-Prot: P43405. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5958