Learn More
Biomatik Corporation Fyn (Phospho-Tyr530) anti-Human, Murine, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41107
The Fyn (Phospho-Tyr530) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine. Immunogen: Synthetic peptide, Human: Fyn around the phosphorylation site of Tyr530. Applications: WB, IHC, ELISA. Gene ID: 2534. Gene Name: FYN. Alternative Names: FYN; P59-FYN; SLK; SYN; kinase Fyn. Swiss-Prot: P06241. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5960