Learn More
Biomatik Corporation Ku70 (Phospho-Ser5) anti-Human, Murine, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41125
The Ku70 (Phospho-Ser5) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine. Immunogen: Synthetic peptide, Human: Ku70 around the phosphorylation site of Ser5. Applications: WB, IHC, ELISA. Gene ID: 2547. Gene Name: XRCC6. Alternative Names: X-ray repair cross-complementing protein 6; 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; X-ray repair complementing defective repair in Chinese hamster cells 6; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subu. Swiss-Prot: P12956. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5963