Learn More
Biomatik Corporation ETS1 (Phospho-Thr38) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41147
The ETS1 (Phospho-Thr38) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: ETS1 around the phosphorylation site of Thr38. Applications: WB, IHC, IF, ELISA. Gene ID: 2113. Gene Name: ETS1. Alternative Names: C-ets-1 protein; ETS-1; ETS1; p54. Swiss-Prot: P14921. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5964