Learn More
Biomatik Corporation IRF-3 (Phospho-Ser385) anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41162
The IRF-3 (Phospho-Ser385) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Synthetic peptide, Human: IRF-3 around the phosphorylation site of Ser385. Applications: WB, IHC, IF, ELISA. Gene ID: 3661. Gene Name: IRF3. Alternative Names: Interferon regulatory factor 3; IRF3. Swiss-Prot: Q14653. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5965