Learn More
Biomatik Corporation MEF2D (Phospho-Ser444) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41175
The MEF2D (Phospho-Ser444) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: MEF2D around the phosphorylation site of Ser444. Applications: WB, IHC, ELISA. Gene ID: 4209. Gene Name: MEF2D. Alternative Names: MEFD; Myocyte-specific enhancer factor 2D. Swiss-Prot: Q14814. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5967