Learn More
Biomatik Corporation Mst1/2 (Phospho-Thr183) anti-Human, Murine, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41284
The Mst1/2 (Phospho-Thr183) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine. Immunogen: Synthetic peptide, Human: Mst1/2 around the phosphorylation site of Thr183. Applications: WB, IHC, ELISA. Gene ID: 6789. Gene Name: STK3. Alternative Names: STE20-like kinase MST1; Yeast Sps1/Ste20-related kinase 3. Swiss-Prot: Q13188/Q13043. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5980