Learn More
Biomatik Corporation DNA-PK (Phospho-Ser2056) anti-Human, Murine, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41427
The DNA-PK (Phospho-Ser2056) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine. Immunogen: Synthetic peptide, Human: DNA-PK around the phosphorylation site of Ser2056. Applications: IHC, ELISA. Gene ID: 5591. Gene Name: PRKDC. Alternative Names: DNA- PKcs; DNA-dependent protein kinase catalytic subunit; DNPK1; P460; PRKD; PRKDC; XRCC7; kinase DNA-PK. Swiss-Prot: P78527. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5987