Learn More
Biomatik Corporation Smad2/3 (Phospho-Thr8) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41478
The Smad2/3 (Phospho-Thr8) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: Smad2/3 around the phosphorylation site of Thr8. Applications: WB, ELISA. Gene ID: 4088/4087. Gene Name: SMAD3. Alternative Names: JV15-2; MAD-3; MADH3; Mad3; Mothers against DPP homolog 3; Mothers against decapentaplegic homolog 3; SMAD 3; Smad 3. Swiss-Prot: P84022/Q15796. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5992