Learn More
Biomatik Corporation RAD51 (Phospho-Tyr315) anti-Human, Murine, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41555
The RAD51 (Phospho-Tyr315) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine. Immunogen: Synthetic peptide, Human: RAD51 around the phosphorylation site of Tyr315. Applications: WB, ELISA. Gene ID: 5888. Gene Name: RAD51. Alternative Names: DNA repair protein RAD51 homolog 1; RAD51A; RECA. Swiss-Prot: Q06609. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5997