Learn More
Biomatik Corporation p47 phox (Phospho-Ser370) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41586
The p47 phox (Phospho-Ser370) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: p47 phox around the phosphorylation site of Ser370. Applications: IHC, ELISA. Gene ID: 653361. Gene Name: NCF1. Alternative Names: 47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NCF-1; NCF-47K; NCF1; Neutrophil cytosol factor 1; Neutrophil cytosolic factor 1; Neutrophil NADPH oxidase factor 1; P47 phox; p47-phox. Swiss-Prot: P14598. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-5999