Learn More
Biomatik Corporation eIF2 alpha (Phospho-Ser51) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41692
The eIF2 alpha (Phospho-Ser51) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: eIF2 alpha around the phosphorylation site of Ser51. Applications: WB, ELISA. Gene ID: 1965. Gene Name: EIF2S1. Alternative Names: eIF-2-alpha; EIF-2A; EIF-2alpha; EIF2S1; Eukaryotic translation initiation factor 2 alpha subunit; Eukaryotic translation initiation factor 2 subunit 1; IF2A. Swiss-Prot: P05198. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6009