Learn More
Biomatik Corporation CASP3 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07002
The CASP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Caspase-3 protein (29-175AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:50-1:200, IF:1:50-1:500. Alternative Names: Apopain, Cysteine protease CPP32Protein Yama, SREBP cleavage activity 1. UniProt: P42574. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6589