Learn More
Biomatik Corporation TP53 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07006
The TP53 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cellular tumor antigen p53 protein (2-393AA). Applications: ELISA, WB, IHC, ChIP. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Antigen NY-CO-13, Phosphoprotein p53, Tumor suppressor p53, TP53, P53. UniProt: P04637. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6593