Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Biomatik Corporation VEGFC anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07016
The VEGFC Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Vascular endothelial growth factor C protein (112-227AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. UniProt: P49767. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6603