Learn More
Biomatik Corporation VEGFA anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07023
The VEGFA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Vascular endothelial growth factor A protein (27-232AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: Vascular endothelial growth factor A, VEGF-A, VEGFA, VEGF, Vascular permeability factor, VPF. UniProt: P15692. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6610