Learn More
Biomatik Corporation HDAC1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07026
Encompass_Preferred
The HDAC1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Histone deacetylase 1 protein (1-482AA). Applications: ELISA, IHC, IF, ChIP. Recommended dilution: IHC:1:20-1:200, IF:1:100-1:500. Alternative Names: Histone deacetylase 1, HD1, HDAC1, RPD3L1. UniProt: Q13547. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6613