Learn More
Biomatik Corporation MAPK1 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07028
The MAPK1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Mitogen-activated protein kinase 1 protein (310-360AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:50-1:500. Alternative Names: Mitogen-activated protein kinase 1, ERT1, Extracellular signal-regulated kinase 2, ERK-2, MAP kinase isoform p42, p42-MAPK, Mitogen-activated protein kinase, MAPK1, ERK2, PRKM1, PRKM2. UniProt: P28482. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6615