Learn More
Biomatik Corporation IkappaB-alpha (Phospho-Ser32/Ser36) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41750
The IkappaB-alpha (Phospho-Ser32/Ser36) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: IkappaB-alpha around the phosphorylation site of Ser32/Ser36. Applications: WB, IHC, IF, ELISA. Gene ID: 4792. Gene Name: NFKBIA. Alternative Names: I-kappa-B-alpha; IkappaBalpha; IKBA; MAD3; Major histocompatibility complex enhancer-binding protein MAD3; NF-kappaB inhibitor alpha; NFKBI; NFKBIA; RL/IF-1. Swiss-Prot: P25963. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6015