Learn More
Biomatik Corporation ATG3 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07035
The ATG3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ubiquitin-like-conjugating enzyme ATG3 protein (1-314AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:1000-1:2000, IF:1:50-1:500. Alternative Names: Autophagy-related protein 3, APG3-like, hApg3, Protein PC3-96, ATG3, APG3, APG3L. UniProt: Q9NT62. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6622