Learn More
Biomatik Corporation mTOR (Phospho-Ser2448) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41782
The mTOR (Phospho-Ser2448) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: mTOR around the phosphorylation site of Ser2448. Applications: WB, IHC, ELISA. Gene ID: 2475. Gene Name: MTOR. Alternative Names: FRAP; FRAP1; FRAP2; RAFT1; Rapamycin target protein; kinase mTOR. Swiss-Prot: P42345. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6016