Learn More
Biomatik Corporation NF-kappaB p65 (Phospho-Ser276) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41795
The NF-kappaB p65 (Phospho-Ser276) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: NF-kappaB p65 around the phosphorylation site of Ser276. Applications: WB, IHC, IP, ELISA. Gene ID: 5970. Gene Name: RELA. Alternative Names: NFKB3; nuclear factor NF-kappa-B p65 subunit; p65; RELA; TF65; transcription factor p65. Swiss-Prot: Q04206. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6017