Learn More
Biomatik Corporation TIMP2 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07054
The TIMP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Metalloproteinase inhibitor 2 protein (30-220AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. UniProt: P16035. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6641