Learn More
Biomatik Corporation CDK2 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07055
The CDK2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cyclin-dependent kinase 2 protein (1-298AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB:1:1000-1:5000, IHC:1:500-1:1000, IF:1:50-1:500, IP:1:200-1:2000. Alternative Names: Cyclin-dependent kinase 2, Cell division protein kinase 2, p33 protein kinase, CDK2, CDKN2. UniProt: P24941. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6642