Learn More
Biomatik Corporation BDNF anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07061
The BDNF Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Brain-derived neurotrophic factor protein (137-237AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Abrineurin, BDNF. UniProt: P23560. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6648