Learn More
Biomatik Corporation CCL5 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07076
The CCL5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human C-C motif chemokine 5 protein (24-91AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. UniProt: P13501. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6663