Learn More
Biomatik Corporation IL2 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07079
The IL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Interleukin-2 protein (21-153AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Interleukin-2, IL-2, IL2, T-cell growth factor, TCGF, INN: Aldesleukin. UniProt: P60568. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6666